Anti-PUMA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX109675
89355-996EA 881.1 CAD
89355-996
Anti-PUMA Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PUMA (BCL2 binding component 3)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PUMA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR