Anti-QKI Rabbit Polyclonal Antibody

GTX30909
89367-700EA 692.7 CAD
89367-700
Anti-QKI Rabbit Polyclonal Antibody
QKI belongs to a family of RNA-binding proteins that have an HNRNPK (MIM 600712) homology (KH) domain embedded in a 200-amino acid region called the GSG domain. Other members of this family include SAM68 (KHDRBS1; MIM 602489) and SF1 (MIM 601516) (Chen and Richard, 1998 [PubMed 9671495]). QKI proteins regulate RNA splicing, export of target RNAs from the nucleus, translation of proteins, and RNA stability (Lauriat et al., 2008 [PubMed 17918747]).[supplied by OMIM]

Type: Primary
Antigen: QKI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR