Anti-RAB11A Rabbit Polyclonal Antibody

GTX113301
89357-450EA 595.28 CAD
89357-450
Anti-RAB11A Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to RAB11A (RAB11A, member RAS oncogene family)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RAB11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR