Anti-Rabbit IgA Goat Polyclonal Antibody

Supplier: GeneTex
GTX77138
89353-514EA 239.78 CAD
89353-514
Anti-Rabbit IgA Goat Polyclonal Antibody
Antibodies
By ELISA, immunoelectrophoresis and double diffusion, the affinity purified antibodies are alpha chain specific.Recommended starting dilution for ELISA: 1/10,000.Recommended starting dilution for Blotting: 1:1000.. Goats were immunized with highly purified rabbit IgA isolated from normal rabbit serum. The resulting anti-IgA was solid phase absorbed and immunoaffinity purified off a rabbit IgA immunoabsorbant

Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR