Anti-RAD21 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX20992
89360-934EA 505.38 CAD
89360-934
Anti-RAD21 Rabbit Polyclonal Antibody
The protein encoded by the Rad21 gene is highly similar to the gene product of Schizosaccharomyces pombe rad21, a gene involved in the repair of DNA double-strand breaks, as well as in chromatid cohesion during mitosis.

Type: Primary
Antigen: RAD21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR