Anti-RAF1 Rat Monoclonal Antibody [clone: pS338 (DPR/2/2A/3A)]

Supplier: GeneTex
GTX75448
89367-878EA 704.54 CAD
89367-878
Anti-RAF1 Rat Monoclonal Antibody [clone: pS338 (DPR/2/2A/3A)]
Antibodies
Rat monoclonal antibody [pS338 (DPR/2/2A/3A)] to Raf1 (phospho Ser338)

Recommended Starting Dilutions: Western Blotting: 1/100 - 1/1000 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: RAF1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR