Anti-RASGRP2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX108616
89355-626EA 778.49 CAD
89355-626
Anti-RASGRP2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to RASGRP2 (RAS guanyl releasing protein 2 (calcium and DAG-regulated))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RASGRP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR