Anti-IgG Mouse Monoclonal Antibody [clone: MARG2c-5] (FITC (Fluorescein Isothiocyanate))

Supplier: GeneTex
GTX11677
89358-706EA 371.04 CAD
89358-706
Anti-IgG Mouse Monoclonal Antibody [clone: MARG2c-5] (FITC (Fluorescein Isothiocyanate))
Antibodies
Rat gamma 2c heavy chain (determined by immunodot). The specificity of monoclonal antibodies against IgG subclasses is determined in a range of optimal concentrations. Increasing the concentration of the first or second antibody can induce cross-reactions with other subclasses.

Recommended Starting Dilutions:FACS: Use at a concentration of 5 µg/mL.Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR