Anti-IgM Goat Polyclonal Antibody

Supplier: GeneTex
GTX77095
89353-490EA 319.98 CAD
89353-490
Anti-IgM Goat Polyclonal Antibody
Antibodies
Goat anti-Rat IgM Polyclonal antibody

Works in most applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR