Anti-RB1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX24783
89361-502EA 497.16 CAD
89361-502
Anti-RB1 Rabbit Polyclonal Antibody
Rabbit polyclonal to RB1 phospho S807, S811

WB: Use at a concentration of 0.25-1.0 ?g/ml. Predicted molecular weight: 10^6 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: RB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR