Anti-RCV1 Mouse Monoclonal Antibody

GTX30629
89367-680EA 574.77 CAD
89367-680
Anti-RCV1 Mouse Monoclonal Antibody
Mouse monoclonal antibody to Recoverin

GTX30629 is useful for immunofluorescence. This antibody may be useful for Western blot on recombinant human recoverin protein where a band of 23 kDa is seen. No other applications have been tested.Suggested working dilutions: *Western blot: 1:500-1:1,000 (recombinant, Alkaline Phosphatase)Immunofluorescence: 1:100-1:500 *The investigator should determine the optimal working dilution for a specific application.

Type: Primary
Antigen: RCV1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR