Anti-RARB Mouse Monoclonal Antibody [clone: 336]

Supplier: GeneTex
GTX30168
89362-878EA 808.49 CAD
89362-878
Anti-RARB Mouse Monoclonal Antibody [clone: 336]
Antibodies
Mouse monoclonal antibody [336] to Retinoic Acid Receptor, ß-Isotype

Type: Primary
Antigen: RARB
Clonality: Monoclonal
Clone: 336
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR