Anti-RFP Mouse Monoclonal Antibody

Supplier: GeneTex

GTX82561
89366-728EA 641.06 CAD
89366-728
Anti-RFP Mouse Monoclonal Antibody
Antibodies
ince the molecular cloning of GFP cDNA and demonstration of GFP as a functional transgene, GFP has become a powerful tool with exciting applications in developmental, cell and molecular biology. RFP is the recent discovered protein that has the similar function and applications as GFP. Either GFP or RFP fluorescence is not species specific and can be expressed in bacteria, yeast, plant and mammalian cells. GFP or RFP can fuse with proteins of interest without interfering significantly with their assembly and function.

Type: Primary
Antigen: RFP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR