Anti-RGS2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX113349
89357-488EA 691.89 CAD
89357-488
Anti-RGS2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to RGS2 (regulator of G-protein signaling 2, 24kDa)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RGS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR