Anti-RGS9 Chicken Polyclonal Antibody

Supplier: Genetex
GTX17444
89367-484EA 1047.45 CAD
89367-484
Anti-RGS9 Chicken Polyclonal Antibody
Antibodies
Members of the RGS family are signaling proteins that suppress the activity of G proteins by promoting their deactivation. Isoforms of RGS9, generated by alternative splicing, show distinct regional and cellular distributions.

For WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: RGS9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR