Anti-RIT2 Rabbit Polyclonal Antibody

GTX104953
89354-042EA 258.54 CAD
89354-042
Anti-RIT2 Rabbit Polyclonal Antibody
RIN belongs to the RAS (HRAS; MIM 190020) superfamily of small GTPases (Shao et al., 1999 [PubMed 10545207]).[supplied by OMIM]

Type: Primary
Antigen: RIT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR