Anti-RKIP Rabbit Monoclonal Antibody

GTX62036
89364-282EA 585.93 CAD
89364-282
Anti-RKIP Rabbit Monoclonal Antibody
Raf kinase inhibitory protein (RKIP) belongs to the phosphatidylethanolamine-binding protein (PEBP) family. It is an inhibitor of the Raf/MEK/MAP kinase signaling cascade and is a suppressor of cancer metastasis . RKIP is involved in neural development, cardiac function and spermatogenesis and appears to have serine protease activity. The inhibition Raf-1-mediated phosphorylation of MEK results from RKIP binding to Raf-1. Dysregulated RKIP expression has the potential to contribute to pathophysiological processes including Alzheimer's disease and diabetic nephropathy . RKIP represents a novel effector of signal transduction pathways leading to apoptosis and a prognostic marker of the pathogenesis of human cancer cells and tumors .

Type: Primary
Antigen: RKIP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR