Anti-RNA Polymerase II RPB11 Mouse Monoclonal Antibody

Supplier: GeneTex
GTX12090
89359-174EA 690.5 CAD
89359-174
Anti-RNA Polymerase II RPB11 Mouse Monoclonal Antibody
Antibodies
RNA polymerase II is responsible for synthesizing messenger RNA in eukaryotes. The product of the RPB11 gene exists as a heterodimer with another polymerase subunit; together they form a core subassembly unit of the polymerase.

Type: Primary
Antigen: RNA Polymerase II RPB11
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR