Anti-RSL1D1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX88161
89366-956EA 767.3 CAD
89366-956
Anti-RSL1D1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to RSL1D1

Approx 75kDa band observed in lysates of cell line K562 (calculated MW of 55.0kDa according to NP_056474.2). The observed molecular weight corresponds to earlier findings with different antibodies from other commercial sources. Recommended concentration: 0.3-1µg/ml.

Type: Primary
Antigen: RSL1D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR