Anti-RT1B Mouse Monoclonal Antibody

Supplier: GeneTex

GTX76188
89366-010EA 640.76 CAD
89366-010
Anti-RT1B Mouse Monoclonal Antibody
Antibodies
Mouse monoclonal [MRC OX-6] to RT1B (Biotin)

For FACS: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: RT1B
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR