Anti-S. Aureus Rabbit Polyclonal Antibody

Supplier: Genetex
GTX40920
89363-362EA 516.58 CAD
89363-362
Anti-S. Aureus Rabbit Polyclonal Antibody
Staphylococcus aureus bacteria can live harmlessly on many skin surfaces, especially around the nose, mouth, genitals, and rectum. But when the skin is punctured or broken for any reason, staphlococcus aureus bacteria can enter the wound and cause an infection.

Type: Primary
Antigen: S. Aureus
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR