Anti-S. Pneumoniae Guinea Pig Polyclonal Antibody

Supplier: GeneTex

GTX40560
89363-228EA 592.37 CAD
89363-228
Anti-S. Pneumoniae Guinea Pig Polyclonal Antibody
Antibodies
Bronchial pneumonia is most prevalant in infants, young children and aged adults. It is caused by various bacteria, including Streptococcus pneumoniae. Streptococcus pneumoniae are Gram-positive, lancet-shaped cocci (elongated cocci with a slightly pointed outer curvature). Usually they are seen as pairs of cocci (diplococci), but they may also occur singly and in short chains.

Type: Primary
Antigen: S. Pneumoniae
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Guinea pig
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR