Anti-SAH3 Rabbit Polyclonal Antibody

GTX11166
89350-798EA 606.37 CAD
89350-798
Anti-SAH3 Rabbit Polyclonal Antibody
In mammals, S Adenosylhomocysteine hydrolase (SAH3 / AdoHcyase) catalyzes the hydrolysis of S adenosylhomocysteine to form adenosine and homocysteine. AdoHcy is the product of all adenosylmethionine (AdoMet) dependent biological Transmethylations. These reactions have a wide range of products, and are common in all facets of biometabolism. As a product inhibitor, elevated levels of AdoHcy suppress AdoMet dependent transmethylations. Thus, SAH3 is a regulator of biological transmethylation.

Type: Primary
Antigen: SAH3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR