Anti-PLK4 Goat Polyclonal Antibody

Supplier: GeneTex
GTX22642
89367-546EA 931.02 CAD
89367-546
Anti-PLK4 Goat Polyclonal Antibody
Antibodies
PLK4 is involved in regulation of cell cycle.

Approx 110kDa band observed in Human Colon lysates (calculated MW of 109kDa according to NP_055079.3). Recommended concentration: 1-3µg/ml.

Type: Primary
Antigen: PLK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR