Anti-SALIVARY Amylase Sheep Polyclonal Antibody

GTX28944
89352-556EA 621.75 CAD
89352-556
Anti-SALIVARY Amylase Sheep Polyclonal Antibody
Salivary Amylase is a digestive enzyme secreted by salivary glands; it consists of a single polypeptide chain with a molecular weight of 56kDa (family B) or 62kDa (family A), the difference in molecular weight being due to the presence or absence of a carbohydrate moiety. The total serum concentration of amylase is about 120g/L of which about 50% is salivary amylase. Although salivary amylase and pancreatic amylase share 95% amino acid sequence homology, they differ in molecular size, isoelectric point and antigenic properties.

Type: Primary
Antigen: SALIVARY Amylase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR