Anti-Salmonella A, B, D Mouse Monoclonal Antibody [clone: 10D9H]

Supplier: GeneTex
GTX28272
89352-384EA 996.89 CAD
89352-384
Anti-Salmonella A, B, D Mouse Monoclonal Antibody [clone: 10D9H]
Antibodies
This antibody detects Salmonella enteriditis D, paratyphi A and typhimurium B

Type: Primary
Antigen: Salmonella A, B, D
Clonality: Monoclonal
Clone: 10D9H
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR