Anti-Salmonella Enteriditis Mouse Monoclonal Antibody

Supplier: GeneTex
GTX28271
89352-382EA 502.11 CAD
89352-382
Anti-Salmonella Enteriditis Mouse Monoclonal Antibody
Antibodies
This antibody detects Salmonella enteriditis D.

Type: Primary
Antigen: Salmonella Enteriditis
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR