Anti-ACT1 Mouse Monoclonal Antibody

Supplier: Genetex
GTX72011
89352-922EA 455.88 CAD
89352-922
Anti-ACT1 Mouse Monoclonal Antibody
This antibody reacts with alpha-skeletal and alpha-cardiac muscle actins. It does not react with smooth muscle.This antibody recognizes skeletal and cardiac muscle tissue.

Type: Primary
Antigen: ACT1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Carp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR