Anti-SELENBP1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX80973
89366-706EA 721.47 CAD
89366-706
Anti-SELENBP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to SELENBP1 (C-term)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500FACS: Use at 1:10-50Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: SELENBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR