Anti-SEPT3 Goat Polyclonal Antibody

GTX22518
89367-544EA 850.86 CAD
89367-544
Anti-SEPT3 Goat Polyclonal Antibody
This gene belongs to the septin family of GTPases. Members of this family are required for cytokinesis. Expression is upregulated by retinoic acid in a human teratocarcinoma cell line. The specific function of this gene has not been determined. Alternative splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq]

Western Blot - Approx 40kDa band seen in Human Brain lysate. Recommended for use at 0.5-2.0ug/ml.

Type: Primary
Antigen: SEPT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR