Anti-Serum Amyloid A Mouse Monoclonal Antibody

GTX20655
89351-526EA 480.99 CAD
89351-526
Anti-Serum Amyloid A Mouse Monoclonal Antibody
Amyloidosis is a heterogeneous group of disorders characterized by extracellular deposition of abnormal protein fibrils, which are derived from different proteins. Almost all patients with amyloidosis have some level of renal disorders. The immunoperoxidase technique has the advantage of being able to detect amyloid in formalin-fixed paraffin embedded tissues and is a very specific method for identifying amyloid fibril proteins. Small and minute amounts of amyloid can be detected with both Amyloid P (GT20688) and Amyloid A antibodies, and thus could allow earlier treatment before organ damage has occurred.

Type: Primary
Antigen: Serum Amyloid A
Clonality: Monoclonal
Clone: "ERROR:#N/A"
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR