Anti-SETDB1 Rabbit Polyclonal Antibody

GTX12317
89350-848EA 504.35 CAD
89350-848
Anti-SETDB1 Rabbit Polyclonal Antibody
Histone methyltransferase methylation represents a specific tag for epigenetic transcriptional repression by recruiting HP1 proteins to methylated histones. Targeted to histone by TRIM28/TIF1B and contributes to silencing of euchromatic genes by KRAB zinc-finger proteins.

Type: Primary
Antigen: SETDB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR