Anti-SF9 Cells Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX17474
89360-312EA 580.76 CAD
89360-312
Anti-SF9 Cells Rabbit Polyclonal Antibody
Antibodies
Cell line from Spodoptera frugiperda (fall armyworm).

Type: Primary
Antigen: SF9 Cells
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR