Anti-SIRPA Mouse Monoclonal Antibody [clone: MRC OX-41]

Supplier: GeneTex

GTX29295
89362-810EA 547.37 CAD
89362-810
Anti-SIRPA Mouse Monoclonal Antibody [clone: MRC OX-41]
Antibodies
Mouse monoclonal [MRC OX - 41 ] to SIRP

Flow Cytometry: 1:100Immunohistochemistry (frozen): 1:20 - 1:100Immunohistochemistry (paraffin): 1:20 - 1:100Flow cytometry: Use 10?l of the suggested working dilution to label 10^6 cells. Where this antibody has not been tested for use in a particular technique this does not necessarily exclude its use in such procedures. Suggested working dilutions are given as a guide only. It is recommended that the user titrates the antibody for use in their own system using appropriate negative/positive controls. This product requires protein digestion pre-treatment of paraffin sections e.g. trypsin or pronase.

Type: Primary
Antigen: SIRPA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR