Anti-SIRT4 Goat Polyclonal Antibody

Supplier: Genetex
GTX10140
89349-014EA 923.94 CAD
89349-014
Anti-SIRT4 Goat Polyclonal Antibody
Antibodies
Goat polyclonal to SIRT4

Peptide ELISA: antibody detection limit dilution >1/32000. WB: Use at an assay dependent dilution. Detects a band of approximately 40 kDa (predicted molecular weight: 36 kDa according to NP_036372) in human kidney and U937 lysates. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SIRT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR