Anti-SLC12A2 Rabbit Polyclonal Antibody

GTX107304
89354-906EA 272.33 CAD
89354-906
Anti-SLC12A2 Rabbit Polyclonal Antibody
By moving chloride into epithelial cells, the Na-K-Cl cotransporter SLC12A2 aids transcellular movement of chloride across both secretory and absorptive epithelia (Payne et al., 1995 [PubMed 7629105]). See also SLC12A1 (MIM 600839) and SLC12A3 (MIM 600968).[supplied by OMIM]

Type: Primary
Antigen: SLC12A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR