Anti-SLC29A1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX85884
89366-898EA 702.96 CAD
89366-898
Anti-SLC29A1 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to SLC29A1

WB (1:50 - 1:100), ELISA (1:1000), IHC-P (10 µg/ml)

Type: Primary
Antigen: SLC29A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR