Anti-SMAD7 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX59874
89363-750EA 768.8 CAD
89363-750
Anti-SMAD7 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Smad7

Western blotting (0.5-4 µg/ml). However, the optimal concentrations should be determined individually. Other applications have not been determined. The antibody detects ~46 kDa Smad-7 from samples of human, mouse, and rat origins. Reactivity to other species has not been determined.

Type: Primary
Antigen: SMAD7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR