Anti-SMC1A Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113299
89357-446EA 881.1 CAD
89357-446
Anti-SMC1A Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SMC1A (structural maintenance of chromosomes protein 1A)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SMC1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR