Anti-SMC2 Rabbit Polyclonal Antibody

GTX10411
89350-764EA 548.04 CAD
89350-764
Anti-SMC2 Rabbit Polyclonal Antibody
Condensin, one of the most abundant components of mitotic chromosomes, is a conserved protein complex composed of two structural maintenance of chromosomes (SMC)1 subunits (SMC2 and SMC4 type) and three non-SMC subunits, and it plays an essential role in mitotic chromosome condensation.

Type: Primary
Antigen: SMC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR