Anti-SMURF2 Rabbit Polyclonal Antibody

GTX110487
89356-318EA 472.59 CAD
89356-318
Anti-SMURF2 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to SMURF2 (SMAD specific E3 ubiquitin protein ligase 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SMURF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR