Anti-Sodium Iodide Symporter Mouse Monoclonal Antibody

GTX17950
89351-432EA 521.39 CAD
89351-432
Anti-Sodium Iodide Symporter Mouse Monoclonal Antibody
Human Sodium Iodide Symporter (hNIS) is responsible for iodide concentrating ability within thyroid follicular cells. It is a membrane bound glycoprotein with 13 membrane spanning domains and 14 extramembranous domains. It may represent an autoantigen in thyroid.

Type: Primary
Antigen: Sodium Iodide Symporter
Clonality: Monoclonal
Clone: "ERROR:#N/A"
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR