Anti-SOX2 Rabbit Monoclonal Antibody

GTX62242
89364-366EA 556.26 CAD
89364-366
Anti-SOX2 Rabbit Monoclonal Antibody
SOX2 is a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. It is required for stem-cell maintenance in the central nervous system, and also regulates gene expression in the stomach . SOX2 is necessary for regulating multiple transcription factors that affect Oct3/4 expression. An essential function of SOX2 is to stabilize embryonic stem cells in a pluripotent state by maintaining the requisite level of Oct3/4 expression .

Type: Primary
Antigen: SOX2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR