Anti-SOX5 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX46041
89363-588EA 888.66 CAD
89363-588
Anti-SOX5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to SOX5

WB: Use at a concentration of 0.2-1 ug/ml. Predicted molecular weight: 84 kDa.

Type: Primary
Antigen: SOX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR