Anti-SRC Rabbit Monoclonal Antibody

GTX61220
89363-884EA 556.26 CAD
89363-884
Anti-SRC Rabbit Monoclonal Antibody
Src is a protein tyrosine kinase known to regulate cellular adhesion. Composed of three major domains; SH2, SH3 and a kinase catalytic domain, Src can be switched from an inactive to an active state though control of its phosphorylation state or through protein-protein interaction. Phosphorylation of Y529 by Csk and Chk inactivates Src, while dephosphorylation of Y529 will modify its conformation to an open active state . Mutation of Y529 leads to Src overactivity. Several cancers including colon and breast cancer have been associated with an increase of Src activity .

Type: Primary
Antigen: SRC
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR