Anti-SRP72 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX102609
89349-662EA 272.33 CAD
89349-662
Anti-SRP72 Rabbit Polyclonal Antibody
The SRP72 gene encodes the 72-kD subunit of the signal recognition particle (SRP), a ribonucleoprotein complex that mediates the targeting of proteins to the endoplasmic reticulum (ER) (summarized by Lutcke et al., 1993 [PubMed 8388879]). The complex consists of a 7S (or 7SL) RNA and 6 different proteins, SRP9 (MIM 600707), SRP14 (MIM 600708), SRP19 (MIM 182175), SRP54 (MIM 604857), SRP68 (MIM 604858), and SRP72. The proteins are bound to the 7S RNA as monomers (SRP19 and SRP54) or heterodimers (SRP9/SRP14 and SRP68/SRP72). SRP9 and SRP14 constitute the Alu domain of 7S, whereas the other 4 proteins belong to the S domain. SRP has at least 3 distinct functions that can be associated with the protein subunits: signal recognition, translational arrest, and ER membrane targeting by interaction with the docking protein.[supplied by OMIM]

Type: Primary
Antigen: SRP72
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR