Anti-TRIM21 Goat Polyclonal Antibody

Supplier: GeneTex
GTX24369
89361-444EA 578.55 CAD
89361-444
Anti-TRIM21 Goat Polyclonal Antibody
Goat polyclonal to SSA1

ELISA: peptide ELISA (antibody detection limit dilution >1 / 32000). WB: Approx. 40 kDa band observed in Human Heart lysate at 0.5?g of primary. Please note that the band we observe is different from the predicted size of approx. 55 kDa according to NP_003132. However, alternatively spliced product of Ro/SSA with a predicted MW of 45 kDa has been observed by Chan et al. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TRIM21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR