Anti-SSBP1 Chicken Polyclonal Antibody

Supplier: Genetex
GTX82400
89368-076EA 721.76 CAD
89368-076
Anti-SSBP1 Chicken Polyclonal Antibody
Chicken polyclonal to Single-stranded DNA-binding protein. mitochondrial

WB (only tested on fusion protein): Use at a dilution of 1:2000. Colorimetric method for signal development.

Type: Primary
Antigen: SSBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR