Anti-STAT1 Rabbit Monoclonal Antibody

Supplier: Genetex
GTX62368
89364-416EA 585.93 CAD
89364-416
Anti-STAT1 Rabbit Monoclonal Antibody
Stat-1 is a latent transcription factor that functions both as a signal transducer in the cytoplasm and an activator of transcription in the nucleus. Stat-1 is one of seven members of the Signal transducer and activators of transcription (STAT) family . Stat-1 is activated by a large number of ligands: IFN-alpha, IFN-gamma, EGF, PDGF and IL6 . IFN-gamma signaling induces phosphorylation of the Stat-1 residue Tyrosine 701, which facilitates dimerization, nuclear translocation, and DNA binding . Upon tyrosine phosphorylation, Stat-1 dimerizes with Stat-2 and associates with ISGF3/IR9 to form a complex called the ISGF3 transcription factor . This activation subsequently allows the complex to translocate to the nucleus and bind specific enhancer elements. Additional complexes can form upon IFN stimulation . Defects in Stat-1 are the cause of Stat-1 deficiency, characterized by mycobacterial or viral diseases .

Type: Primary
Antigen: STAT1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR