Anti-STK3 Mouse Monoclonal Antibody [clone: 4G10]

Supplier: GeneTex
GTX83546
89366-774EA 1067.02 CAD
89366-774
Anti-STK3 Mouse Monoclonal Antibody [clone: 4G10]
Antibodies
Mouse monoclonal antibody [4G10] to STK3

Recommended Starting Dilutions:WB 1:2000; IHC 1:50Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: STK3
Clonality: Monoclonal
Clone: 4G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR